Learn More
Meso Scale Discovery U-PLEX Human S100A12 ASY
SDP
Catalog No. 5000079050
Encompass
The Meso Scale Discovery U-PLEX Human S100A12 Assay is a multiplex immunoassay that measures S100A12 protein levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can be fully automated on the MSD instrument platforms The U-PLEX format allows simultaneous measurement of S100A12 with other biomarkers in human samples Overall this is a sensitive reproducible assay for quantifying S100A12 levels in various human sample types
Catalog No. 50-000-79050 Supplier Meso Scale Discovery Supplier No. K151AHPK2
The Meso Scale Discovery U-PLEX Human S100A12 Assay is a multiplex immunoassay that measures S100A12 protein levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can be fully automated on the MSD instrument platforms The U-PLEX format allows simultaneous measurement of S100A12 with other biomarkers in human samples Overall this is a sensitive reproducible assay for quantifying S100A12 levels in various human sample types