Learn More
Meso Scale Discovery U-PLEX Human S100A12 ASY
SDP
Catalog No. 5000079559
Encompass
The Meso Scale Discovery U-PLEX Human S100A12 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of S100A12 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of S100A12 an inflammatory protein biomarker This assay has high sensitivity a broad dynamic range and requires a low sample volume It provides reproducible and quantitative data for research use including biomarker discovery translational research and drug development
Catalog No. 50-000-79559 Supplier Meso Scale Discovery Supplier No. K151AHPK1
The Meso Scale Discovery U-PLEX Human S100A12 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of S100A12 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of S100A12 an inflammatory protein biomarker This assay has high sensitivity a broad dynamic range and requires a low sample volume It provides reproducible and quantitative data for research use including biomarker discovery translational research and drug development