Learn More
Meso Scale Discovery U-PLEX Human SAA ASY 25 PL
SDP
Catalog No. 5000082229
Encompass
The Meso Scale Discovery U-PLEX Human SAA Assay Kit is an electrochemiluminescent immunoassay for measuring serum amyloid A (SAA) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls diluents wash buffer read buffer and plate seals Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL
Catalog No. 50-000-82229 Supplier Meso Scale Discovery Supplier No. K151F9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human SAA Assay Kit is an electrochemiluminescent immunoassay for measuring serum amyloid A (SAA) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls diluents wash buffer read buffer and plate seals Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL