Learn More
Meso Scale Discovery U-PLEX Human SAA ASY
SDP
Catalog No. 5000078337
Encompass
The Meso Scale Discovery U-PLEX Human SAA Assay Kit is an electrochemiluminescent immunoassay for measuring serum amyloid A (SAA) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantitation of SAA levels between 0 3 - 1000 ng/mL The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents SAA is an acute phase protein that increases during inflammation making the assay useful for monitoring inflammatory diseases
Catalog No. 50-000-78337 Supplier Meso Scale Discovery Supplier No. K151F9K2
The Meso Scale Discovery U-PLEX Human SAA Assay Kit is an electrochemiluminescent immunoassay for measuring serum amyloid A (SAA) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantitation of SAA levels between 0 3 - 1000 ng/mL The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents SAA is an acute phase protein that increases during inflammation making the assay useful for monitoring inflammatory diseases