Learn More
Meso Scale Discovery U-PLEX Human SAA ASY384-well
SDP
Catalog No. 5000080687
Encompass
The Meso Scale Discovery U-PLEX Human SAA 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of serum amyloid A (SAA) in human serum plasma and cell culture supernatants The assay has a detection range of 0 3-1000 ng/mL and requires a sample volume of 25 L The total assay time is approximately 3 5 hours This multiplex assay allows high-throughput screening and provides sensitive accurate and reproducible results using Meso Scale Discovery instrumentation and 384-well plates
Catalog No. 50-000-80687 Supplier Meso Scale Discovery Supplier No. K251F9K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human SAA 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of serum amyloid A (SAA) in human serum plasma and cell culture supernatants The assay has a detection range of 0 3-1000 ng/mL and requires a sample volume of 25 L The total assay time is approximately 3 5 hours This multiplex assay allows high-throughput screening and provides sensitive accurate and reproducible results using Meso Scale Discovery instrumentation and 384-well plates