Learn More
Meso Scale Discovery U-PLEX Human SAA ASYQP
SDP
Catalog No. 5000078535
Encompass
The Meso Scale Discovery U-PLEX Human SAA Assay Kit is an electrochemiluminescent immunoassay for measuring serum amyloid A (SAA) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantification of SAA levels from 0 3 - 1000 ng/mL The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent It performs up to 80 tests in about 3 5 hours offering a simple rapid and sensitive method for quantifying SAA in human samples
Catalog No. 50-000-78535 Supplier Meso Scale Discovery Supplier No. K151F9K21
The Meso Scale Discovery U-PLEX Human SAA Assay Kit is an electrochemiluminescent immunoassay for measuring serum amyloid A (SAA) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantification of SAA levels from 0 3 - 1000 ng/mL The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent It performs up to 80 tests in about 3 5 hours offering a simple rapid and sensitive method for quantifying SAA in human samples