Learn More
Meso Scale Discovery U-PLEX Human SAA ASYQP
SDP
Catalog No. 5000081985
Encompass
The Meso Scale Discovery U-PLEX Human SAA Assay Kit is an electrochemiluminescent immunoassay for quantifying serum amyloid A (SAA) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and necessary reagents Results are analyzed on the MESO QuickPlex SQ 120 The assay quantifies from 0 3 ng/mL to 1500 ng/mL
Catalog No. 50-000-81985 Supplier Meso Scale Discovery Supplier No. K151F9K22
The Meso Scale Discovery U-PLEX Human SAA Assay Kit is an electrochemiluminescent immunoassay for quantifying serum amyloid A (SAA) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and necessary reagents Results are analyzed on the MESO QuickPlex SQ 120 The assay quantifies from 0 3 ng/mL to 1500 ng/mL