Learn More
Meso Scale Discovery U-PLEX Human SDF-1 ASY
SDP
Catalog No. 5000081431
Encompass
The Meso Scale Discovery U-PLEX Human SDF-1alpha Assay Kit is a multiplex immunoassay that measures SDF-1alpha levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrators and reagents needed to run the assay Results are obtained in as little as 3 5 hours This assay allows for the quantitative measurement of SDF-1alpha levels for life science research applications
Catalog No. 50-000-81431 Supplier Meso Scale Discovery Supplier No. K151VBK1
The Meso Scale Discovery U-PLEX Human SDF-1alpha Assay Kit is a multiplex immunoassay that measures SDF-1alpha levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrators and reagents needed to run the assay Results are obtained in as little as 3 5 hours This assay allows for the quantitative measurement of SDF-1alpha levels for life science research applications