Learn More
Meso Scale Discovery U-PLEX Human SDF-1 ASYQP
SDP
Catalog No. 5000079756
Encompass
The MSD U-PLEX Human SDF-1 Singleplex Assay is an electrochemiluminescence-based immunoassay kit designed for quantitative measurement of stromal cell-derived factor-1 alpha (SDF-1/CXCL12) in human biological samples This singleplex format allows focused analysis of this important chemokine involved in cell migration hematopoiesis and immune responses The assay utilizes Meso Scale Discovery s proprietary U-PLEX platform technology offering high sensitivity wide dynamic range and minimal sample volume requirements for research applications
Catalog No. 50-000-79756 Supplier Meso Scale Discovery Supplier No. K151VBK22
Add to Cart
Add to Cart
The MSD U-PLEX Human SDF-1 Singleplex Assay is an electrochemiluminescence-based immunoassay kit designed for quantitative measurement of stromal cell-derived factor-1 alpha (SDF-1/CXCL12) in human biological samples This singleplex format allows focused analysis of this important chemokine involved in cell migration hematopoiesis and immune responses The assay utilizes Meso Scale Discovery s proprietary U-PLEX platform technology offering high sensitivity wide dynamic range and minimal sample volume requirements for research applications