Learn More
Meso Scale Discovery U-PLEX Human Serpin A1 ASY
SDP
Catalog No. 5000078727
Encompass
The Meso Scale Discovery U-PLEX Human Serpin A1 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Serpin A1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L or less sample volume and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This assay provides a simple rapid way to measure Serpin A1 levels across multiple samples
This item is not returnable. View return policy
Catalog No. 50-000-78727 Supplier Meso Scale Discovery Supplier No. K151W9K1
The Meso Scale Discovery U-PLEX Human Serpin A1 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Serpin A1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L or less sample volume and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This assay provides a simple rapid way to measure Serpin A1 levels across multiple samples