Learn More
Meso Scale Discovery U-PLEX Human SHBG ASY 25 PL
SDP
Catalog No. 5000082466
Encompass
The Meso Scale Discovery U-PLEX Human SHBG Assay Kit measures Sex Hormone Binding Globulin (SHBG) in human serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of SHBG levels down to 0 2 ng/mL The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls buffers and seals It is compatible with MSD s SECTOR Imager 6000 for high throughput screening enabling reliable SHBG quantification in biological samples
Catalog No. 50-000-82466 Supplier Meso Scale Discovery Supplier No. K151G9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human SHBG Assay Kit measures Sex Hormone Binding Globulin (SHBG) in human serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of SHBG levels down to 0 2 ng/mL The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls buffers and seals It is compatible with MSD s SECTOR Imager 6000 for high throughput screening enabling reliable SHBG quantification in biological samples