Learn More
Meso Scale Discovery U-PLEX Human SHBG ASY
SDP
Catalog No. 5000081612
Encompass
The Meso Scale Discovery U-PLEX Human SHBG Assay Kit measures SHBG in serum plasma and cell culture supernatants using electrochemiluminescence (ECL) technology It offers high sensitivity broad dynamic range and precision The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators and controls Sensitivity is 0 08 nmol/L with a quantification limit of 200 nmol/L Results are available in 3 5 hours using the MSD QuickPlex SQ 120 Instrument enabling accurate SHBG quantification for research
Catalog No. 50-000-81612 Supplier Meso Scale Discovery Supplier No. K151G9K1
The Meso Scale Discovery U-PLEX Human SHBG Assay Kit measures SHBG in serum plasma and cell culture supernatants using electrochemiluminescence (ECL) technology It offers high sensitivity broad dynamic range and precision The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators and controls Sensitivity is 0 08 nmol/L with a quantification limit of 200 nmol/L Results are available in 3 5 hours using the MSD QuickPlex SQ 120 Instrument enabling accurate SHBG quantification for research