Learn More
Meso Scale Discovery U-PLEX Human SHBG ASY
SDP
Catalog No. 5000082011
Encompass
The Meso Scale Discovery U-PLEX Human SHBG Assay Kit is a multiplex immunoassay that measures human sex hormone-binding globulin (SHBG) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can quantify SHBG concentrations between 0 05-1000 ng/mL It includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X read buffer 10X wash buffer and substrate The kit has high lot-to-lot consistency and takes approximately 3 5 hours to complete
Catalog No. 50-000-82011 Supplier Meso Scale Discovery Supplier No. K151G9K2
The Meso Scale Discovery U-PLEX Human SHBG Assay Kit is a multiplex immunoassay that measures human sex hormone-binding globulin (SHBG) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can quantify SHBG concentrations between 0 05-1000 ng/mL It includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X read buffer 10X wash buffer and substrate The kit has high lot-to-lot consistency and takes approximately 3 5 hours to complete