Learn More
Meso Scale Discovery U-PLEX Human SHBG ASY384-well
SDP
Catalog No. 5000079858
Encompass
The Meso Scale Discovery U-PLEX Human SHBG 384-Well Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of sex hormone-binding globulin (SHBG) in human serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and buffers for 384 tests The assay has a typical sensitivity of 0 2 nmol/L and a detection range of 0 2-200 nmol/L Results are obtained in under 3 hours using an MSD instrument capable of reading electrochemiluminescent signals This assay allows high-throughput sensitive quantification of SHBG levels
Catalog No. 50-000-79858 Supplier Meso Scale Discovery Supplier No. K251G9K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human SHBG 384-Well Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of sex hormone-binding globulin (SHBG) in human serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and buffers for 384 tests The assay has a typical sensitivity of 0 2 nmol/L and a detection range of 0 2-200 nmol/L Results are obtained in under 3 hours using an MSD instrument capable of reading electrochemiluminescent signals This assay allows high-throughput sensitive quantification of SHBG levels