Learn More
Meso Scale Discovery U-PLEX Human SHBG ASYQP
SDP
Catalog No. 5000080188
Encompass
The Meso Scale Discovery U-PLEX Human SHBG Assay Kit is a multiplex immunoassay that quantitatively measures human sex hormone-binding globulin (SHBG) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls The assay has a typical sensitivity of 0 05 ng/mL and requires 25 L sample volume per well
Catalog No. 50-000-80188 Supplier Meso Scale Discovery Supplier No. K151G9K22
The Meso Scale Discovery U-PLEX Human SHBG Assay Kit is a multiplex immunoassay that quantitatively measures human sex hormone-binding globulin (SHBG) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls The assay has a typical sensitivity of 0 05 ng/mL and requires 25 L sample volume per well