Learn More
Meso Scale Discovery U-PLEX Human SHBG ASYQP
SDP
Catalog No. 5000081614
Encompass
The Meso Scale Discovery U-PLEX Human SHBG Assay Kit is an electrochemiluminescent immunoassay for measuring sex hormone-binding globulin (SHBG) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a 96-well plate detection antibody calibrators and necessary reagents Sensitivity is 0 05 nmol/L with a range of 0 05 to 50 nmol/L The assay takes about 3 5 hours for 96 tests enabling high-throughput SHBG analysis in research
Catalog No. 50-000-81614 Supplier Meso Scale Discovery Supplier No. K151G9K21
The Meso Scale Discovery U-PLEX Human SHBG Assay Kit is an electrochemiluminescent immunoassay for measuring sex hormone-binding globulin (SHBG) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a 96-well plate detection antibody calibrators and necessary reagents Sensitivity is 0 05 nmol/L with a range of 0 05 to 50 nmol/L The assay takes about 3 5 hours for 96 tests enabling high-throughput SHBG analysis in research