Learn More
Meso Scale Discovery U-PLEX Human sTfR-1 ASYQP
SDP
Catalog No. 5000080581
Encompass
The Meso Scale Discovery U-PLEX Human STFR-1 Assay is a multiplex immunoassay kit for measuring soluble transferrin receptor 1 (sTFR1) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs This assay accurately quantifies sTFR1 indicating iron deficiency and erythropoiesis aiding in diagnosing anemia and inflammatory diseases With minimal hands-on time it is a simple rapid tool for sTFR1 detection in clinical research
Catalog No. 50-000-80581 Supplier Meso Scale Discovery Supplier No. K151P9K21
The Meso Scale Discovery U-PLEX Human STFR-1 Assay is a multiplex immunoassay kit for measuring soluble transferrin receptor 1 (sTFR1) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs This assay accurately quantifies sTFR1 indicating iron deficiency and erythropoiesis aiding in diagnosing anemia and inflammatory diseases With minimal hands-on time it is a simple rapid tool for sTFR1 detection in clinical research