Learn More
Meso Scale Discovery U-PLEX Human sTfR-1 ASYQP
SDP
Catalog No. 5000081261
Encompass
The U-PLEX Human STFR-1 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures soluble transferrin receptor 1 (sTfR1) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay allows for the quantitative measurement of sTfR1 which can be used to evaluate iron deficiency and erythropoiesis The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and control
Catalog No. 50-000-81261 Supplier Meso Scale Discovery Supplier No. K151P9K22
The U-PLEX Human STFR-1 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures soluble transferrin receptor 1 (sTfR1) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay allows for the quantitative measurement of sTfR1 which can be used to evaluate iron deficiency and erythropoiesis The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and control