Learn More
Meso Scale Discovery U-PLEX Human TARC ASY
SDP
Catalog No. 5000078241
Encompass
The Meso Scale Discovery U-PLEX Human TARC Assay is a multiplex immunoassay kit for measuring Thymus and Activation Regulated Chemokine (TARC/CCL17) in human serum plasma and cell culture supernatant It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture and detection antibodies calibrators and controls It runs on MSD s SECTOR Imager platform with data analysis via MSD Discovery Workbench software providing a sensitive and reproducible method for quantifying TARC levels
Catalog No. 50-000-78241 Supplier Meso Scale Discovery Supplier No. K151ULK1
The Meso Scale Discovery U-PLEX Human TARC Assay is a multiplex immunoassay kit for measuring Thymus and Activation Regulated Chemokine (TARC/CCL17) in human serum plasma and cell culture supernatant It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture and detection antibodies calibrators and controls It runs on MSD s SECTOR Imager platform with data analysis via MSD Discovery Workbench software providing a sensitive and reproducible method for quantifying TARC levels