Learn More
Meso Scale Discovery U-PLEX Human TARC ASYQP
SDP
Catalog No. 5000079036
Encompass
The U-PLEX Human TARC Assay (K151ULK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of TARC (Thymus and Activation-Regulated Chemokine/CCL17) in human biological samples This electrochemiluminescence-based assay utilizes U-PLEX technology for sensitive specific detection with minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and controls for reliable reproducible results in research applications studying inflammatory responses and immune cell recruitment
Catalog No. 50-000-79036 Supplier Meso Scale Discovery Supplier No. K151ULK22
The U-PLEX Human TARC Assay (K151ULK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of TARC (Thymus and Activation-Regulated Chemokine/CCL17) in human biological samples This electrochemiluminescence-based assay utilizes U-PLEX technology for sensitive specific detection with minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and controls for reliable reproducible results in research applications studying inflammatory responses and immune cell recruitment