Learn More
Meso Scale Discovery U-PLEX Human TGF-B1 ASYQP
SDP
Catalog No. 5000079667
Encompass
The U-PLEX Human TGF 1 Assay (K151XWK) is a multiplex electrochemiluminescence immunoassay from Meso Scale Discovery for quantitative measurement of transforming growth factor beta 1 in human biological samples This research-grade assay utilizes proprietary MULTI-SPOT and MULTI-ARRAY technology platforms offering high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable TGF 1 quantification in serum plasma and cell culture supernatants for biomedical research applications
Catalog No. 50-000-79667 Supplier Meso Scale Discovery Supplier No. K151XWK22
The U-PLEX Human TGF 1 Assay (K151XWK) is a multiplex electrochemiluminescence immunoassay from Meso Scale Discovery for quantitative measurement of transforming growth factor beta 1 in human biological samples This research-grade assay utilizes proprietary MULTI-SPOT and MULTI-ARRAY technology platforms offering high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable TGF 1 quantification in serum plasma and cell culture supernatants for biomedical research applications