Learn More
Meso Scale Discovery U-PLEX Human TGF-B2 ASY
SDP
Catalog No. 5000080094
Encompass
The Meso Scale Discovery U-PLEX Human TGF-beta2 Assay Kit is designed for the quantitative measurement of active and total TGF-beta2 in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and buffers for sample dilution and washing The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for a sample volume of 25 L The kit can perform up to 80 tests in duplicate and has a testing time of 3 5 hours
Catalog No. 50-000-80094 Supplier Meso Scale Discovery Supplier No. K151XUK1
The Meso Scale Discovery U-PLEX Human TGF-beta2 Assay Kit is designed for the quantitative measurement of active and total TGF-beta2 in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and buffers for sample dilution and washing The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for a sample volume of 25 L The kit can perform up to 80 tests in duplicate and has a testing time of 3 5 hours