Learn More
Meso Scale Discovery U-PLEX Human TGF-B2 ASYQP
SDP
Catalog No. 5000079818
Encompass
The Meso Scale Discovery U-PLEX Human TGF-beta2 Assay Kit is designed for the quantitative measurement of active and total TGF-beta2 in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD s proprietary QuickPlex SQ 120 instrument and provides high sensitivity and a broad dynamic range This multiplex kit includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and quantify sample concentrations The assay has a lower limit of detection of 0 06 pg/mL and an upper limit of quantification of 10 000 pg/mL for measuring TGF-beta2
Catalog No. 50-000-79818 Supplier Meso Scale Discovery Supplier No. K151XUK22
The Meso Scale Discovery U-PLEX Human TGF-beta2 Assay Kit is designed for the quantitative measurement of active and total TGF-beta2 in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD s proprietary QuickPlex SQ 120 instrument and provides high sensitivity and a broad dynamic range This multiplex kit includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and quantify sample concentrations The assay has a lower limit of detection of 0 06 pg/mL and an upper limit of quantification of 10 000 pg/mL for measuring TGF-beta2