Learn More
Meso Scale Discovery U-PLEX Human TGF-B3 ASYQP
SDP
Catalog No. 5000079243
Encompass
The Meso Scale Discovery U-PLEX Human TGF-beta3 Assay Kit is designed for the quantitative measurement of active and total TGF-beta3 in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD s proprietary SULFO-TAG labels and multi-array/multi-spot plate technology to provide sensitive and rapid detection of TGF-beta3 levels This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrator diluent wash buffer read buffer and a 10-point standard curve for quantitative analysis of samples ranging from 0 61 pg/mL to 10 000 pg/mL
Catalog No. 50-000-79243 Supplier Meso Scale Discovery Supplier No. K151XVK21
The Meso Scale Discovery U-PLEX Human TGF-beta3 Assay Kit is designed for the quantitative measurement of active and total TGF-beta3 in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD s proprietary SULFO-TAG labels and multi-array/multi-spot plate technology to provide sensitive and rapid detection of TGF-beta3 levels This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrator diluent wash buffer read buffer and a 10-point standard curve for quantitative analysis of samples ranging from 0 61 pg/mL to 10 000 pg/mL