Learn More
Meso Scale Discovery U-PLEX Human Tie-2 Assay, Single-analyte, 96-well plate, ECL detection, Dynamic range 2.9 - 29000 pg/mL, Sample types: Plasma, Serum, Capture Antibody: Mouse Monoclonal, Detection Antibody: Goat Polyclonal
SDP
Catalog No. NC3793878
Encompass
U-PLEX Human Tie-2 Assay, Single-analyte, 96-well plate, ECL detection, Dynamic range 2.9 - 29000 pg/mL, Sample types: Plasma, Serum, Capture Antibody: Mouse Monoclonal, Detection Antibody: Goat Polyclonal
Catalog No. NC3793878 Supplier Meso Scale Discovery Supplier No. K151F8K2
U-PLEX Human Tie-2 Assay, Single-analyte, 96-well plate, ECL detection, Dynamic range 2.9 - 29000 pg/mL, Sample types: Plasma, Serum, Capture Antibody: Mouse Monoclonal, Detection Antibody: Goat Polyclonal