Learn More
Meso Scale Discovery U-PLEX Human Tie-2 ASY 25 PL
SDP
Catalog No. 5000081655
Encompass
The Meso Scale Discovery U-PLEX Human TIE-2 Assay Kit is designed for the quantitative measurement of human TIE-2 in serum plasma and cell culture supernatants using an electrochemiluminescent detection method The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent The assay has a typical sensitivity of 0 3 ng/mL and an upper limit of quantification of 2500 pg/mL This immunoassay allows high-throughput sensitive and accurate quantification of human TIE-2 levels
Catalog No. 50-000-81655 Supplier Meso Scale Discovery Supplier No. K151F8K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TIE-2 Assay Kit is designed for the quantitative measurement of human TIE-2 in serum plasma and cell culture supernatants using an electrochemiluminescent detection method The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent The assay has a typical sensitivity of 0 3 ng/mL and an upper limit of quantification of 2500 pg/mL This immunoassay allows high-throughput sensitive and accurate quantification of human TIE-2 levels