Learn More
Meso Scale Discovery U-PLEX Human Tie-2 ASY
SDP
Catalog No. 5000082228
Encompass
The Meso Scale Discovery U-PLEX Human TIE-2 Assay Kit is designed for the quantitative measurement of human TIE-2 in serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates to provide superior sensitivity and a wide dynamic range This assay allows for the precise and reproducible measurement of human TIE-2 with a lower limit of detection of 19 5 pg/mL and an upper limit of 100 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator
Catalog No. 50-000-82228 Supplier Meso Scale Discovery Supplier No. K151F8K1
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Human TIE-2 Assay Kit is designed for the quantitative measurement of human TIE-2 in serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates to provide superior sensitivity and a wide dynamic range This assay allows for the precise and reproducible measurement of human TIE-2 with a lower limit of detection of 19 5 pg/mL and an upper limit of 100 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator