Learn More
Meso Scale Discovery U-PLEX Human Tie-2 ASY384-well
SDP
Catalog No. 5000081047
Encompass
The Meso Scale Discovery U-PLEX Human TIE-2 384-Well Assay Kit is a multiplex immunoassay that measures human TIE-2 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control samples It allows simultaneous measurement of TIE-2 levels across 384 samples in approximately 5 hours The kit has high performance and reliability for biomarker research and drug discovery applications
Catalog No. 50-000-81047 Supplier Meso Scale Discovery Supplier No. K251F8K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TIE-2 384-Well Assay Kit is a multiplex immunoassay that measures human TIE-2 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control samples It allows simultaneous measurement of TIE-2 levels across 384 samples in approximately 5 hours The kit has high performance and reliability for biomarker research and drug discovery applications