Learn More
Meso Scale Discovery U-PLEX Human Tie-2 ASYQP
SDP
Catalog No. 5000082036
Encompass
The Meso Scale Discovery U-PLEX Human TIE-2 Assay Kit is a multiplex immunoassay that measures human TIE-2 in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and diluent The assay takes approximately 3 5 hours to run and only requires 10 L of sample
Catalog No. 50-000-82036 Supplier Meso Scale Discovery Supplier No. K151F8K22
The Meso Scale Discovery U-PLEX Human TIE-2 Assay Kit is a multiplex immunoassay that measures human TIE-2 in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and diluent The assay takes approximately 3 5 hours to run and only requires 10 L of sample