Learn More
Meso Scale Discovery U-PLEX Human TIGIT ASY 25 PL
SDP
Catalog No. 5000082365
Encompass
The Meso Scale Discovery U-PLEX Human TIGIT Assay Kit is a multiplex immunoassay for quantifying human TIGIT and other biomarkers in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results from small samples The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators controls diluents wash buffer read buffer and plate seals The assay s sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL for TIGIT
Catalog No. 50-000-82365 Supplier Meso Scale Discovery Supplier No. K151W7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TIGIT Assay Kit is a multiplex immunoassay for quantifying human TIGIT and other biomarkers in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results from small samples The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators controls diluents wash buffer read buffer and plate seals The assay s sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL for TIGIT