Learn More
Meso Scale Discovery U-PLEX Human TIGIT ASY
SDP
Catalog No. 5000078765
Encompass
The Meso Scale Discovery U-PLEX Human TIGIT Assay Kit is a multiplex immunoassay for quantifying human TIGIT and other biomarkers in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results from small samples The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators controls diluents wash buffer read buffer and seals Sensitivity is 0 3 pg/mL with an upper quantification limit of 2500 pg/mL for TIGIT
Catalog No. 50-000-78765 Supplier Meso Scale Discovery Supplier No. K151W7K1
The Meso Scale Discovery U-PLEX Human TIGIT Assay Kit is a multiplex immunoassay for quantifying human TIGIT and other biomarkers in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results from small samples The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators controls diluents wash buffer read buffer and seals Sensitivity is 0 3 pg/mL with an upper quantification limit of 2500 pg/mL for TIGIT