Learn More
Meso Scale Discovery U-PLEX Human TIGIT ASY384-well
SDP
Catalog No. 5000080652
Encompass
The Meso Scale Discovery U-PLEX Human TIGIT 384-Well Assay is a multiplex immunoassay that enables simultaneous quantification of human TIGIT and other biomarkers in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is compatible with high-throughput screening platforms The kit includes a 384-well plate coated with capture antibodies detection antibodies calibrators and controls needed to perform the assay Results are obtained in as little as 3 5 hours with high sensitivity and broad dynamic range This assay provides a rapid reproducible method for profiling TIGIT and other immune checkpoint proteins
Catalog No. 50-000-80652 Supplier Meso Scale Discovery Supplier No. K251W7K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TIGIT 384-Well Assay is a multiplex immunoassay that enables simultaneous quantification of human TIGIT and other biomarkers in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is compatible with high-throughput screening platforms The kit includes a 384-well plate coated with capture antibodies detection antibodies calibrators and controls needed to perform the assay Results are obtained in as little as 3 5 hours with high sensitivity and broad dynamic range This assay provides a rapid reproducible method for profiling TIGIT and other immune checkpoint proteins