Learn More
Meso Scale Discovery U-PLEX Human TIGIT ASYQP
SDP
Catalog No. 5000082364
Encompass
The Meso Scale Discovery U-PLEX Human TIGIT Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human TIGIT and other biomarkers in serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies Read Buffer Calibrator Blend and Sample Diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for TIGIT
Catalog No. 50-000-82364 Supplier Meso Scale Discovery Supplier No. K151W7K22
The Meso Scale Discovery U-PLEX Human TIGIT Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human TIGIT and other biomarkers in serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies Read Buffer Calibrator Blend and Sample Diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for TIGIT