Learn More
Meso Scale Discovery U-PLEX Human TLR1 ASYQP
SDP
Catalog No. 5000079275
Encompass
The U-PLEX Human TLR1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human Toll-like receptor 1 (TLR1) in cell and tissue lysates This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and minimal sample consumption The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag and calibrators and controls to generate a standard curve and validate assay performance The assay measures TLR1 levels quantitatively with high precision and accuracy It is suitable for studying TLR1 biology and immune response
Catalog No. 50-000-79275 Supplier Meso Scale Discovery Supplier No. K151J7K21
The U-PLEX Human TLR1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human Toll-like receptor 1 (TLR1) in cell and tissue lysates This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and minimal sample consumption The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag and calibrators and controls to generate a standard curve and validate assay performance The assay measures TLR1 levels quantitatively with high precision and accuracy It is suitable for studying TLR1 biology and immune response