Learn More
Meso Scale Discovery U-PLEX Human TNF ASYQP
SDP
Catalog No. 5000080505
Encompass
The U-PLEX Human TNF Assay (K151UCK) is a highly sensitive immunoassay kit from Meso Scale Discovery for quantitative measurement of tumor necrosis factor-alpha in human biological samples This multiplex-compatible assay utilizes electrochemiluminescence detection technology to provide superior sensitivity and dynamic range compared to traditional ELISA methods The kit includes pre-coated plates detection antibodies and calibration standards for reliable reproducible results in research applications studying inflammation immune response and cytokine signaling pathways
Catalog No. 50-000-80505 Supplier Meso Scale Discovery Supplier No. K151UCK22
The U-PLEX Human TNF Assay (K151UCK) is a highly sensitive immunoassay kit from Meso Scale Discovery for quantitative measurement of tumor necrosis factor-alpha in human biological samples This multiplex-compatible assay utilizes electrochemiluminescence detection technology to provide superior sensitivity and dynamic range compared to traditional ELISA methods The kit includes pre-coated plates detection antibodies and calibration standards for reliable reproducible results in research applications studying inflammation immune response and cytokine signaling pathways