Learn More
Meso Scale Discovery U-PLEX Human TNF-B ASY 25 PL
SDP
Catalog No. 5000080579
Encompass
The Meso Scale Discovery U-PLEX Human TNF-beta Assay Kit is a multiplex immunoassay that measures human TNF-beta in serum plasma cell culture supernatants and other biological samples It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can be run on Meso Scale Discovery instrumentation including the QuickPlex SQ 120 QuickPlex SQ 120i MESO QuickPlex SQ 120 and V-PLEX Plus The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and ancillary reagents needed to perform the assay
Catalog No. 50-000-80579 Supplier Meso Scale Discovery Supplier No. K151UUK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TNF-beta Assay Kit is a multiplex immunoassay that measures human TNF-beta in serum plasma cell culture supernatants and other biological samples It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can be run on Meso Scale Discovery instrumentation including the QuickPlex SQ 120 QuickPlex SQ 120i MESO QuickPlex SQ 120 and V-PLEX Plus The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and ancillary reagents needed to perform the assay