Learn More
Meso Scale Discovery U-PLEX Human TNF-B ASY
SDP
Catalog No. 5000079703
Encompass
The Meso Scale Discovery U-PLEX Human TNF-beta Assay Kit is a multiplex immunoassay that measures TNF-beta levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay has high specificity and low cross-reactivity It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and buffer The kit allows simultaneous measurement of TNF-beta and other biomarkers It can be used for biomarker discovery lead optimization and diagnostics
Catalog No. 50-000-79703 Supplier Meso Scale Discovery Supplier No. K151UUK2
The Meso Scale Discovery U-PLEX Human TNF-beta Assay Kit is a multiplex immunoassay that measures TNF-beta levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay has high specificity and low cross-reactivity It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and buffer The kit allows simultaneous measurement of TNF-beta and other biomarkers It can be used for biomarker discovery lead optimization and diagnostics