Learn More
Meso Scale Discovery U-PLEX Human TNF-B ASY384-well
SDP
Catalog No. 5000080653
Encompass
The Meso Scale Discovery U-PLEX Human TNF-beta 384-well Assay is a multiplex immunoassay kit that enables simultaneous quantification of the cytokine TNF-beta in human serum plasma and cell culture supernatant samples It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can be run in 384-well format for high throughput applications It includes a 10-plate kit with reagents and consumables for 3 840 data points
Catalog No. 50-000-80653 Supplier Meso Scale Discovery Supplier No. K251UUK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TNF-beta 384-well Assay is a multiplex immunoassay kit that enables simultaneous quantification of the cytokine TNF-beta in human serum plasma and cell culture supernatant samples It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can be run in 384-well format for high throughput applications It includes a 10-plate kit with reagents and consumables for 3 840 data points