Learn More
Meso Scale Discovery U-PLEX Human TNF-RI ASYQP
SDP
Catalog No. 5000078407
Encompass
The Meso Scale Discovery U-PLEX Human TNF RI Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human TNF receptor I (TNFR1) in serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls for generating a standard curve and measuring TNFR1 concentrations between 0 05-1500 pg/mL It can monitor soluble TNFR1 levels and TNF pathway activation
Catalog No. 50-000-78407 Supplier Meso Scale Discovery Supplier No. K151AHQK21
The Meso Scale Discovery U-PLEX Human TNF RI Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human TNF receptor I (TNFR1) in serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls for generating a standard curve and measuring TNFR1 concentrations between 0 05-1500 pg/mL It can monitor soluble TNFR1 levels and TNF pathway activation