Learn More
Meso Scale Discovery U-PLEX Human TNF-RII ASYQP
SDP
Catalog No. 5000081592
Encompass
The Meso Scale Discovery U-PLEX Human TNF RII Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human TNF RII in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on Meso Scale Discovery SECTOR Instruments It includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance
Catalog No. 50-000-81592 Supplier Meso Scale Discovery Supplier No. K151AHRK22
The Meso Scale Discovery U-PLEX Human TNF RII Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human TNF RII in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on Meso Scale Discovery SECTOR Instruments It includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance