Learn More
Meso Scale Discovery U-PLEX Human TPO ASY 25 PL
SDP
Catalog No. 5000079593
Encompass
The Meso Scale Discovery U-PLEX Human TPO Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Thyroid Peroxidase (TPO) in human serum plasma and cell culture supernatants This assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours The assay has a lower limit of detection of 0 03 ng/mL and an upper limit of quantification of 1500 ng/mL This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls The U-PLEX platform provides a streamlined and efficient workflow for biomarker quantification
Catalog No. 50-000-79593 Supplier Meso Scale Discovery Supplier No. K151VKK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TPO Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Thyroid Peroxidase (TPO) in human serum plasma and cell culture supernatants This assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours The assay has a lower limit of detection of 0 03 ng/mL and an upper limit of quantification of 1500 ng/mL This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls The U-PLEX platform provides a streamlined and efficient workflow for biomarker quantification