Learn More
Meso Scale Discovery U-PLEX Human TPO ASY
SDP
Catalog No. 5000081956
Encompass
The Meso Scale Discovery U-PLEX Human TPO Assay is an electrochemiluminescent immunoassay for measuring Thyroid Peroxidase (TPO) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results in 3 5 hours The detection range is 0 08 to 2500 ng/mL The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals offering a simple rapid and sensitive method to quantify TPO levels
Catalog No. 50-000-81956 Supplier Meso Scale Discovery Supplier No. K151VKK1
The Meso Scale Discovery U-PLEX Human TPO Assay is an electrochemiluminescent immunoassay for measuring Thyroid Peroxidase (TPO) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results in 3 5 hours The detection range is 0 08 to 2500 ng/mL The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals offering a simple rapid and sensitive method to quantify TPO levels