Learn More
Meso Scale Discovery U-PLEX Human TPO ASY384-well
SDP
Catalog No. 5000080662
Encompass
The Meso Scale Discovery U-PLEX Human TPO 384-Well Assay Kit is a 384-well multiplex immunoassay that measures human Thyroid Peroxidase (TPO) in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well MULTI-ARRAY Plate Detection Antibody Read Buffer Calibrator and Diluent It can test up to 384 samples in a single run and has a testing time of approximately 3 5 hours The kit is for research use only
Catalog No. 50-000-80662 Supplier Meso Scale Discovery Supplier No. K251VKK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TPO 384-Well Assay Kit is a 384-well multiplex immunoassay that measures human Thyroid Peroxidase (TPO) in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well MULTI-ARRAY Plate Detection Antibody Read Buffer Calibrator and Diluent It can test up to 384 samples in a single run and has a testing time of approximately 3 5 hours The kit is for research use only