Learn More
Meso Scale Discovery U-PLEX Human TRAIL ASY
SDP
Catalog No. 5000079629
Encompass
The U-PLEX Human TRAIL Assay K151XTK from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of TRAIL in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a 96-well plate format The kit includes a plate coated with capture antibodies detection antibodies calibrators and diluents The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL for TRAIL The total assay time is approximately 3 5 hours This product allows for the accurate quantitation of TRAIL across a wide dynamic range
Catalog No. 50-000-79629 Supplier Meso Scale Discovery Supplier No. K151XTK2
The U-PLEX Human TRAIL Assay K151XTK from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of TRAIL in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a 96-well plate format The kit includes a plate coated with capture antibodies detection antibodies calibrators and diluents The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL for TRAIL The total assay time is approximately 3 5 hours This product allows for the accurate quantitation of TRAIL across a wide dynamic range