Learn More
Meso Scale Discovery U-PLEX Human TSLP ASY
SDP
Catalog No. 5000081901
Encompass
The Meso Scale Discovery U-PLEX Human TSLP Assay Kit is a multiplex immunoassay that allows simultaneous measurement of human thymic stromal lymphopoietin (TSLP) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and diluents needed to perform the assay TSLP is measured in pg/mL concentrations with an assay range from 0 17 pg/mL to 2500 pg/mL This assay allows high-throughput quantitative analysis of TSLP
Catalog No. 50-000-81901 Supplier Meso Scale Discovery Supplier No. K151WHK1
The Meso Scale Discovery U-PLEX Human TSLP Assay Kit is a multiplex immunoassay that allows simultaneous measurement of human thymic stromal lymphopoietin (TSLP) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and diluents needed to perform the assay TSLP is measured in pg/mL concentrations with an assay range from 0 17 pg/mL to 2500 pg/mL This assay allows high-throughput quantitative analysis of TSLP