Learn More
Meso Scale Discovery U-PLEX Human VCAM-1 ASY
SDP
Catalog No. 5000079274
Encompass
The Meso Scale Discovery U-PLEX Human VCAM-1 Assay Kit is a multiplex immunoassay that measures VCAM-1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Discovery instrumentation including the MESO QuickPlex SQ 120 QuickPlex SQ 120 and V-PLEX Plus The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrators It allows quantification of VCAM-1 levels with a lower limit of detection of 0 02 ng/mL
Catalog No. 50-000-79274 Supplier Meso Scale Discovery Supplier No. K151H9K1
The Meso Scale Discovery U-PLEX Human VCAM-1 Assay Kit is a multiplex immunoassay that measures VCAM-1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Discovery instrumentation including the MESO QuickPlex SQ 120 QuickPlex SQ 120 and V-PLEX Plus The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrators It allows quantification of VCAM-1 levels with a lower limit of detection of 0 02 ng/mL