Learn More
Meso Scale Discovery U-PLEX Human VCAM-1 ASY
SDP
Catalog No. 5000082273
Encompass
The Meso Scale Discovery U-PLEX Human VCAM-1 Assay Kit measures VCAM-1 in human serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of VCAM-1 at sub-pg/mL levels The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators controls diluents buffers and seals The assay takes about 3 5 hours and is compatible with MSD s SECTOR Imager for reading It allows high-throughput quantitative VCAM-1 measurement in human samples
Catalog No. 50-000-82273 Supplier Meso Scale Discovery Supplier No. K151H9K2
The Meso Scale Discovery U-PLEX Human VCAM-1 Assay Kit measures VCAM-1 in human serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of VCAM-1 at sub-pg/mL levels The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators controls diluents buffers and seals The assay takes about 3 5 hours and is compatible with MSD s SECTOR Imager for reading It allows high-throughput quantitative VCAM-1 measurement in human samples