Learn More
Meso Scale Discovery U-PLEX Human VCAM-1 ASYQP
SDP
Catalog No. 5000079271
Encompass
The Meso Scale Discovery U-PLEX Human VCAM-1 Assay Kit is a multiplex immunoassay that measures VCAM-1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can monitor soluble VCAM-1 levels as a biomarker of endothelial dysfunction and inflammation It requires 50 L of sample and can be read on MSD instrument platforms The kit includes a 96-well plate coated with capture antibodies detection antibodies and calibrators ranging from 1 22 pg/mL to 10 000 pg/mL
Catalog No. 50-000-79271 Supplier Meso Scale Discovery Supplier No. K151H9K22
The Meso Scale Discovery U-PLEX Human VCAM-1 Assay Kit is a multiplex immunoassay that measures VCAM-1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can monitor soluble VCAM-1 levels as a biomarker of endothelial dysfunction and inflammation It requires 50 L of sample and can be read on MSD instrument platforms The kit includes a 96-well plate coated with capture antibodies detection antibodies and calibrators ranging from 1 22 pg/mL to 10 000 pg/mL