Learn More
Meso Scale Discovery U-PLEX Human VEGF-a ASY
SDP
Catalog No. 5000079734
Encompass
The Meso Scale Discovery U-PLEX Human VEGF-A Assay Kit is a multiplex immunoassay for measuring VEGF-A in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The assay offers high specificity and low cross-reactivity It includes a plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrator diluent wash buffer read buffer and a 10-point VEGF-A standard curve The kit is customizable with a 3 5-hour testing time suitable for biomarker research drug discovery and clinical research
Catalog No. 50-000-79734 Supplier Meso Scale Discovery Supplier No. K151UVK2
The Meso Scale Discovery U-PLEX Human VEGF-A Assay Kit is a multiplex immunoassay for measuring VEGF-A in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The assay offers high specificity and low cross-reactivity It includes a plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrator diluent wash buffer read buffer and a 10-point VEGF-A standard curve The kit is customizable with a 3 5-hour testing time suitable for biomarker research drug discovery and clinical research