Learn More
Meso Scale Discovery U-PLEX Human VEGF-a ASY
SDP
Catalog No. 5000080431
Encompass
The Meso Scale Discovery U-PLEX Human VEGF-A Assay Kit is a multiplex immunoassay that measures VEGF-A levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay has high specificity and low cross-reactivity It includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrator and buffer The kit allows quantification of VEGF-A levels with a lower limit of detection of 0 06 pg/mL
Catalog No. 50-000-80431 Supplier Meso Scale Discovery Supplier No. K151UVK1
The Meso Scale Discovery U-PLEX Human VEGF-A Assay Kit is a multiplex immunoassay that measures VEGF-A levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay has high specificity and low cross-reactivity It includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrator and buffer The kit allows quantification of VEGF-A levels with a lower limit of detection of 0 06 pg/mL